Assay Details
| Gene Symbol: stag2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100028.1 | NP_001093498.1 | 9 - 10 | 1099 | 110 | |
| XM_009301356.3 | XP_009299631.1 | 9 - 10 | 1160 | 110 | ||
| XM_017356146.2 | XP_017211635.1 | 9 - 10 | 1242 | 110 | ||
More Information
Set Membership: |
|

stag2a
Genomic Map