Assay Details
| Gene Symbol: men1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130953.1 | NP_571028.1 | 9 - 10 | 1446 | 72 | |
| GenBank mRNA | AF212919.1 | - | 9 - 10 | 1446 | 72 | |
| AJ245952.1 | - | 9 - 10 | 1435 | 72 | ||
| BC095301.1 | - | 9 - 10 | 1505 | 72 | ||
More Information
Set Membership: |
|

men1
Genomic Map