Assay Details
| Gene Symbol: dld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130955.2 | NP_571030.2 | 4 - 5 | 1123 | 59 | |
| GenBank mRNA | BC129159.1 | - | 4 - 5 | 1123 | 59 | |
| CU458992.1 | - | 4 - 5 | 671 | 59 | ||
| Y11760.1 | - | 671 | 59 | |||
More Information
Set Membership: |
|

dld
Genomic Map