Assay Details
| Gene Symbol: snai1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313699.1 | NP_001300628.1 | 3 - 4 | 823 | 102 | |
| NM_001313700.1 | NP_001300629.1 | 2 - 3 | 807 | 102 | ||
| GenBank mRNA | BC049300.1 | - | 2 - 3 | 732 | 102 | |
| BC092698.1 | - | 2 - 3 | 637 | 102 | ||
| BC152602.1 | - | 3 - 4 | 824 | 102 | ||
| BC164590.1 | - | 2 - 3 | 625 | 102 | ||
| EH544887.1 | - | 2 - 3 | 810 | 102 | ||
| S68799.1 | - | 2 - 3 | 817 | 102 | ||
| X74790.1 | - | 705 | 102 | |||
More Information
Set Membership: |
|

snai1a
Genomic Map