Assay Details
| Gene Symbol: foxo3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131085.1 | NP_571160.1 | 3 - 4 | 2669 | 157 | |
| GenBank mRNA | AF114262.1 | - | 3 - 4 | 2669 | 157 | |
| BC162481.1 | - | 3 - 4 | 2517 | 157 | ||
More Information
Set Membership: |
|

foxo3b
Genomic Map