Assay Details
| Gene Symbol: hoxc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131123.1 | NP_571198.1 | 1 - 2 | 404 | 78 | |
| GenBank mRNA | BC083307.1 | - | 1 - 2 | 618 | 78 | |
| BC165049.1 | - | 1 - 2 | 369 | 78 | ||
| DQ060554.1 | - | 1 - 2 | 287 | 78 | ||
More Information
Set Membership: |
|

hoxc6a
Genomic Map