Assay Details
| Gene Symbol: nr6a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131256.2 | NP_571331.2 | 5 - 6 | 682 | 82 | |
| XM_017357271.2 | XP_017212760.1 | 4 - 5 | 966 | 82 | ||
| GenBank mRNA | AJ007703.2 | - | 5 - 6 | 682 | 82 | |
| BC162651.1 | - | 4 - 5 | 523 | 82 | ||
More Information
Set Membership: |
|

nr6a1a
Genomic Map