Assay Details
| Gene Symbol: nr6a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131256.2 | NP_571331.2 | 6 - 7 | 899 | 68 | |
| XM_017357271.2 | XP_017212760.1 | 5 - 6 | 1183 | 68 | ||
| GenBank mRNA | AJ007703.2 | - | 6 - 7 | 899 | 68 | |
| BC162651.1 | - | 5 - 6 | 740 | 68 | ||
More Information
Set Membership: |
|

nr6a1a
Genomic Map