Assay Details
| Gene Symbol: nr6a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131256.2 | NP_571331.2 | 9 - 10 | 1428 | 63 | |
| XM_017357271.2 | XP_017212760.1 | 8 - 9 | 1712 | 63 | ||
| GenBank mRNA | AJ007703.2 | - | 9 - 10 | 1428 | 63 | |
| BC162651.1 | - | 8 - 9 | 1269 | 63 | ||
More Information
Set Membership: |
|

nr6a1a
Genomic Map