Assay Details
| Gene Symbol: tp53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271820.1 | NP_001258749.1 | 9 - 10 | 1101 | 93 | |
| NM_001328587.1 | NP_001315516.1 | 9 - 10 | 1086 | 93 | ||
| NM_001328588.1 | NP_001315517.1 | 6 - 7 | 846 | 93 | ||
| NM_131327.2 | NP_571402.1 | 9 - 10 | 1098 | 93 | ||
| XM_005165101.4 | XP_005165158.1 | 9 - 10 | 948 | 93 | ||
| XM_005165103.4 | XP_005165160.1 | 6 - 7 | 974 | 93 | ||
| XM_005165104.4 | XP_005165161.1 | 6 - 7 | 798 | 93 | ||
| GenBank mRNA | BC095597.1 | - | 9 - 10 | 1089 | 93 | |
| CT704385.2 | - | 3 - 4 | 330 | 93 | ||
| KM981741.1 | - | 6 - 7 | 852 | 93 | ||
| KM981742.1 | - | 6 - 7 | 856 | 93 | ||
| KM981743.1 | - | 7 - 8 | 1000 | 93 | ||
| U60804.1 | - | 9 - 10 | 970 | 93 | ||
More Information
Set Membership: |
|

tp53
Genomic Map