Assay Details
| Gene Symbol: cpeb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131427.2 | NP_571502.2 | 2 - 3 | 222 | 75 | |
| XM_021477394.1 | XP_021333069.1 | 2 - 3 | 251 | 75 | ||
| GenBank mRNA | AB011680.1 | - | 1 - 2 | 186 | 75 | |
| AF076918.1 | - | 2 - 3 | 201 | 75 | ||
| BC045467.1 | - | 2 - 3 | 220 | 75 | ||
| BC164038.1 | - | 2 - 3 | 220 | 75 | ||
| EH503358.1 | - | 2 - 3 | 257 | 75 | ||
More Information
Set Membership: |
|

cpeb1b
Genomic Map