Assay Details
| Gene Symbol: cpeb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131427.2 | NP_571502.2 | 5 - 6 | 880 | 62 | |
| XM_021477394.1 | XP_021333069.1 | 5 - 6 | 909 | 62 | ||
| GenBank mRNA | AB011680.1 | - | 4 - 5 | 844 | 62 | |
| AF076918.1 | - | 5 - 6 | 859 | 62 | ||
| BC045467.1 | - | 5 - 6 | 878 | 62 | ||
| BC164038.1 | - | 5 - 6 | 878 | 62 | ||
More Information
Set Membership: |
|

cpeb1b
Genomic Map