Assay Details
| Gene Symbol: cpeb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131427.2 | NP_571502.2 | 6 - 7 | 984 | 112 | |
| XM_021477394.1 | XP_021333069.1 | 6 - 7 | 1013 | 112 | ||
| GenBank mRNA | AB011680.1 | - | 5 - 6 | 948 | 112 | |
| AF076918.1 | - | 6 - 7 | 963 | 112 | ||
| BC045467.1 | - | 6 - 7 | 982 | 112 | ||
| BC164038.1 | - | 6 - 7 | 982 | 112 | ||
More Information
Set Membership: |
|

cpeb1b
Genomic Map