Assay Details
| Gene Symbol: cpeb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131427.2 | NP_571502.2 | 7 - 8 | 1095 | 68 | |
| GenBank mRNA | AB011680.1 | - | 6 - 7 | 1059 | 68 | |
| AF076918.1 | - | 7 - 8 | 1074 | 68 | ||
| BC045467.1 | - | 7 - 8 | 1093 | 68 | ||
| BC164038.1 | - | 7 - 8 | 1093 | 68 | ||
More Information
Set Membership: |
|

cpeb1b
Genomic Map