Assay Details
| Gene Symbol: bmpr1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131457.2 | NP_571532.1 | 13 - 14 | 1767 | 87 | |
| XM_021476002.1 | XP_021331677.1 | 9 - 10 | 1407 | 87 | ||
| GenBank mRNA | AB020758.1 | - | 10 - 11 | 1469 | 87 | |
| BC081625.1 | - | 13 - 14 | 1767 | 87 | ||
More Information
Set Membership: |
|

bmpr1ba
Genomic Map