Assay Details
| Gene Symbol: ctbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131714.2 | NP_571789.2 | 6 - 7 | 773 | 108 | |
| XM_005173133.2 | XP_005173190.1 | 4 - 5 | 730 | 108 | ||
| GenBank mRNA | AB032415.1 | - | 6 - 7 | 826 | 108 | |
| BC045280.1 | - | 6 - 7 | 773 | 108 | ||
More Information
Set Membership: |
|

ctbp1
Genomic Map