Assay Details
| Gene Symbol: cry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131786.2 | NP_571861.2 | 1 - 2 | 265 | 66 | |
| GenBank mRNA | AB042252.1 | - | 1 - 2 | 154 | 66 | |
| BC046088.1 | - | 1 - 2 | 265 | 66 | ||
More Information
Set Membership: |
|

cry2
Genomic Map