Assay Details
| Gene Symbol: cry1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131790.4 | NP_571865.4 | 12 - 13 | 2030 | 76 | |
| GenBank mRNA | BC044558.1 | - | 12 - 13 | 2030 | 76 | |
More Information
Set Membership: |
|

cry1b
Genomic Map