Assay Details
| Gene Symbol: cry1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131790.4 | NP_571865.4 | 3 - 4 | 607 | 129 | |
| GenBank mRNA | AB042249.1 | - | 2 - 3 | 262 | 129 | |
| BC044558.1 | - | 3 - 4 | 607 | 129 | ||
More Information
Set Membership: |
|

cry1b
Genomic Map