Assay Details
| Gene Symbol: cry1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131790.4 | NP_571865.4 | 6 - 7 | 1028 | 97 | |
| GenBank mRNA | AB042249.1 | - | 5 - 6 | 683 | 97 | |
| BC044558.1 | - | 6 - 7 | 1028 | 97 | ||
More Information
Set Membership: |
|

cry1b
Genomic Map