Assay Details
| Gene Symbol: igf1rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152969.1 | NP_694501.1 | 13 - 14 | 2748 | 73 | |
| GenBank mRNA | AF400276.1 | - | 13 - 14 | 2748 | 73 | |
| AY043191.1 | - | 1 - 2 | 155 | 73 | ||
| BC163581.1 | - | 13 - 14 | 2779 | 73 | ||
More Information
Set Membership: |
|

igf1rb
Genomic Map