Assay Details
| Gene Symbol: igf1rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152969.1 | NP_694501.1 | 17 - 18 | 3257 | 76 | |
| GenBank mRNA | AF329815.1 | - | 4 - 5 | 434 | 76 | |
| AF400270.1 | - | 2 - 3 | 245 | 76 | ||
| AF400276.1 | - | 17 - 18 | 3257 | 76 | ||
| AY043191.1 | - | 5 - 6 | 664 | 76 | ||
| BC163581.1 | - | 17 - 18 | 3288 | 76 | ||
More Information
Set Membership: |
|

igf1rb
Genomic Map