Assay Details
| Gene Symbol: igf1rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152969.1 | NP_694501.1 | 6 - 7 | 1420 | 125 | |
| GenBank mRNA | AF400276.1 | - | 6 - 7 | 1420 | 125 | |
| BC163581.1 | - | 6 - 7 | 1451 | 125 | ||
More Information
Set Membership: |
|

igf1rb
Genomic Map