Assay Details
| Gene Symbol: malt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152976.1 | NP_694508.1 | 15 - 16 | 2082 | 130 | |
| XM_005161701.4 | XP_005161758.1 | 15 - 16 | 2164 | 130 | ||
| GenBank mRNA | AF316598.1 | - | 15 - 16 | 2082 | 130 | |
More Information
Set Membership: |
|

malt2
Genomic Map