Assay Details
| Gene Symbol: smarca4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181603.1 | NP_853634.1 | 15 - 16 | 2474 | 71 | |
| XM_021469728.1 | XP_021325403.1 | 16 - 17 | 2661 | 71 | ||
| GenBank mRNA | AY205256.1 | - | 15 - 16 | 2474 | 71 | |
| BC163918.1 | - | 15 - 16 | 2536 | 71 | ||
| BC163920.1 | - | 15 - 16 | 2536 | 71 | ||
More Information
Set Membership: |
|

smarca4a
Genomic Map