Assay Details
| Gene Symbol: tfpia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309803.1 | NP_001296732.1 | 5 - 6 | 725 | 148 | |
| NM_182872.2 | NP_878292.2 | 5 - 6 | 722 | 148 | ||
| GenBank mRNA | AF515274.1 | - | 5 - 6 | 586 | 148 | |
| BC133127.1 | - | 5 - 6 | 639 | 148 | ||
More Information
Set Membership: |
|

tfpia
Genomic Map