Assay Details
| Gene Symbol: mhc1zaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194425.1 | NP_919406.1 | 1173 | 68 | ||
| GenBank mRNA | AJ420953.1 | - | 1173 | 68 | ||
| BC063995.1 | - | 1206 | 68 | |||
| BC076561.1 | - | 1211 | 68 | |||
| BC124481.1 | - | 1208 | 68 | |||
More Information
Set Membership: |
|

mhc1zaa
Genomic Map