Assay Details
| Gene Symbol: rad21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199595.1 | NP_955889.1 | 13 - 14 | 1888 | 60 | |
| GenBank mRNA | AY423040.1 | - | 13 - 14 | 1897 | 60 | |
| BC045311.1 | - | 13 - 14 | 1888 | 60 | ||
| BC164557.1 | - | 12 - 13 | 1777 | 60 | ||
More Information
Set Membership: |
|

rad21a
Genomic Map