Assay Details
| Gene Symbol: rad21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199595.1 | NP_955889.1 | 5 - 6 | 630 | 71 | |
| GenBank mRNA | AY423040.1 | - | 5 - 6 | 639 | 71 | |
| BC045311.1 | - | 5 - 6 | 630 | 71 | ||
| BC164557.1 | - | 4 - 5 | 519 | 71 | ||
More Information
Set Membership: |
|

rad21a
Genomic Map