Assay Details
| Gene Symbol: rad21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199595.1 | NP_955889.1 | 6 - 7 | 823 | 72 | |
| GenBank mRNA | BC045311.1 | - | 6 - 7 | 823 | 72 | |
| BC164557.1 | - | 5 - 6 | 712 | 72 | ||
More Information
Set Membership: |
|

rad21a
Genomic Map