Assay Details
| Gene Symbol: zfp36l1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199649.2 | NP_955943.1 | 1 - 2 | 186 | 121 | |
| GenBank mRNA | BC063991.1 | - | 1 - 2 | 186 | 121 | |
More Information
Set Membership: |
|

zfp36l1b
Genomic Map