Assay Details
| Gene Symbol: ddx39aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328342.1 | NP_001315271.1 | 5 - 6 | 808 | 111 | |
| GenBank mRNA | BC042330.1 | - | 6 - 7 | 722 | 111 | |
| BC071505.1 | - | 5 - 6 | 810 | 111 | ||
| BC164154.1 | - | 3 - 4 | 519 | 111 | ||
| EH433199.1 | - | 5 - 6 | 822 | 111 | ||
| EH559791.1 | - | 5 - 6 | 799 | 111 | ||
More Information
Set Membership: |
|

ddx39aa
Genomic Map