Assay Details
| Gene Symbol: cyth1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199722.1 | NP_956016.1 | 4 - 5 | 421 | 109 | |
| XM_005169466.4 | XP_005169523.1 | 4 - 5 | 490 | 109 | ||
| XM_017358433.2 | XP_017213922.1 | 4 - 5 | 471 | 109 | ||
| GenBank mRNA | BC059497.1 | - | 4 - 5 | 421 | 109 | |
More Information
Set Membership: |
|

cyth1b
Genomic Map