Assay Details
| Gene Symbol: pex26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199920.1 | NP_956214.1 | 2 - 3 | 453 | 65 | |
| GenBank mRNA | BC053199.1 | - | 2 - 3 | 453 | 65 | |
| BC164102.1 | - | 2 - 3 | 387 | 65 | ||
More Information
Set Membership: |
|

pex26
Genomic Map