Assay Details
| Gene Symbol: tsc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200052.1 | NP_956346.1 | 15 - 16 | 2106 | 71 | |
| GenBank mRNA | BC055190.1 | - | 15 - 16 | 2106 | 71 | |
More Information
Set Membership: |
|

tsc1a
Genomic Map