Assay Details
| Gene Symbol: tsc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200052.1 | NP_956346.1 | 19 - 20 | 2693 | 93 | |
| XM_005171795.4 | XP_005171852.1 | 19 - 20 | 2791 | 93 | ||
| XM_005171796.4 | XP_005171853.1 | 19 - 20 | 2801 | 93 | ||
| XM_005171797.4 | XP_005171854.1 | 19 - 20 | 2713 | 93 | ||
| XM_021476019.1 | XP_021331694.1 | 11 - 12 | 2009 | 93 | ||
| GenBank mRNA | BC055190.1 | - | 19 - 20 | 2693 | 93 | |
More Information
Set Membership: |
|

tsc1a
Genomic Map