Assay Details
| Gene Symbol: tsc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200052.1 | NP_956346.1 | 7 - 8 | 895 | 145 | |
| XM_005171795.4 | XP_005171852.1 | 7 - 8 | 966 | 145 | ||
| XM_005171796.4 | XP_005171853.1 | 7 - 8 | 976 | 145 | ||
| XM_005171797.4 | XP_005171854.1 | 7 - 8 | 966 | 145 | ||
| GenBank mRNA | BC055190.1 | - | 7 - 8 | 895 | 145 | |
More Information
Set Membership: |
|

tsc1a
Genomic Map