Assay Details
| Gene Symbol: dcaf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200129.1 | NP_956423.1 | 9 - 10 | 1195 | 63 | |
| GenBank mRNA | BC044155.1 | - | 9 - 10 | 1195 | 63 | |
| BC164738.1 | - | 9 - 10 | 1119 | 63 | ||
More Information
Set Membership: |
|

dcaf13
Genomic Map