Assay Details
| Gene Symbol: zw10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200141.2 | NP_956435.1 | 5 - 6 | 628 | 75 | |
| GenBank mRNA | BC044489.1 | - | 5 - 6 | 633 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

zw10| Gene Symbol: zw10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200141.2 | NP_956435.1 | 5 - 6 | 628 | 75 | |
| GenBank mRNA | BC044489.1 | - | 5 - 6 | 633 | 75 | |
Set Membership: |
|