Assay Details
| Gene Symbol: scpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200192.1 | NP_956486.1 | 1 - 2 | 150 | 65 | |
| GenBank mRNA | BC045898.1 | - | 1 - 2 | 150 | 65 | |
| BC066718.1 | - | 1 - 2 | 109 | 65 | ||
More Information
Set Membership: |
|

scpep1
Genomic Map