Assay Details
| Gene Symbol: tmem110l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200197.1 | NP_956491.1 | 1 - 2 | 373 | 102 | |
| GenBank mRNA | BC045927.1 | - | 1 - 2 | 373 | 102 | |
More Information
Set Membership: |
|

tmem110l
Genomic Map