Assay Details
| Gene Symbol: tmem110l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200197.1 | NP_956491.1 | 7 - 8 | 965 | 70 | |
| XM_009300367.3 | XP_009298642.1 | 8 - 9 | 748 | 70 | ||
| XM_021474968.1 | XP_021330643.1 | 7 - 8 | 621 | 70 | ||
| GenBank mRNA | BC045927.1 | - | 7 - 8 | 965 | 70 | |
More Information
Set Membership: |
|

tmem110l
Genomic Map