Assay Details
| Gene Symbol: erap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200206.2 | NP_956500.2 | 2 - 3 | 633 | 62 | |
| XM_005171800.4 | XP_005171857.1 | 2 - 3 | 650 | 62 | ||
| XM_009301453.3 | XP_009299728.1 | 2 - 3 | 836 | 62 | ||
More Information
Set Membership: |
|

erap1b
Genomic Map