Assay Details
| Gene Symbol: prc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200234.1 | NP_956528.1 | 10 - 11 | 1473 | 61 | |
| GenBank mRNA | BC046886.1 | - | 10 - 11 | 1473 | 61 | |
| BC075764.1 | - | 10 - 11 | 1456 | 61 | ||
More Information
Set Membership: |
|

prc1b
Genomic Map