Assay Details
| Gene Symbol: prc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200234.1 | NP_956528.1 | 12 - 13 | 1688 | 64 | |
| GenBank mRNA | BC046886.1 | - | 12 - 13 | 1688 | 64 | |
| BC075764.1 | - | 12 - 13 | 1671 | 64 | ||
More Information
Set Membership: |
|

prc1b
Genomic Map