Assay Details
| Gene Symbol: rtraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200270.2 | NP_956564.2 | 2 - 3 | 334 | 109 | |
| GenBank mRNA | BC049049.1 | - | 2 - 3 | 295 | 109 | |
| EH547502.1 | - | 2 - 3 | 322 | 109 | ||
More Information
Set Membership: |
|

rtraf
Genomic Map