Assay Details
| Gene Symbol: rtraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200270.2 | NP_956564.2 | 3 - 4 | 438 | 65 | |
| GenBank mRNA | BC049049.1 | - | 3 - 4 | 399 | 65 | |
| EH460317.1 | - | 5 - 6 | 762 | 65 | ||
| EH547502.1 | - | 3 - 4 | 426 | 65 | ||
More Information
Set Membership: |
|

rtraf
Genomic Map