Assay Details
| Gene Symbol: rtraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200270.2 | NP_956564.2 | 5 - 6 | 594 | 123 | |
| GenBank mRNA | BC049049.1 | - | 5 - 6 | 555 | 123 | |
| EH460317.1 | - | 3 - 4 | 606 | 123 | ||
| EH547502.1 | - | 5 - 6 | 582 | 123 | ||
More Information
Set Membership: |
|

rtraf
Genomic Map