Assay Details
| Gene Symbol: znhit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200273.1 | NP_956567.1 | 2 - 3 | 184 | 67 | |
| XM_021476032.1 | XP_021331707.1 | 2 - 3 | 208 | 67 | ||
| GenBank mRNA | BC049063.1 | - | 2 - 3 | 184 | 67 | |
More Information
Set Membership: |
|

znhit3
Genomic Map