Assay Details
| Gene Symbol: pigb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200476.1 | NP_956770.1 | 8 - 9 | 1084 | 67 | |
| GenBank mRNA | BC055191.1 | - | 8 - 9 | 1084 | 67 | |
More Information
Set Membership: |
|

pigb
Genomic Map